3-Brom-N-(furan-2-ylmethyl)-5-(trifluormethyl)benzolsulfonamid

Lieferant: COMBI-BLOCKS

Synonyme: 3-Bromo-5-[N-(fur-2-ylmethyl)sulphamoyl]benzotrifluoride

HC-6295-1G HC-6295-5G
COBBHC-6295-1GEA 0 EUR
COBBHC-6295-1G COBBHC-6295-5G
3-Brom-N-(furan-2-ylmethyl)-5-(trifluormethyl)benzolsulfonamid
3-Brom-N-(furan-2-ylmethyl)-5-(trifluormethyl)benzolsulfonamid
Formel: C₁₂H₉BrF₃NO₃S
MDL: MFCD09801006
CAS-Nummer: 951884-84-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR