Transfection reagents, transfection kit, TransIT®-mRNA
Lieferant: MIRUS
MIR2255 MIR2256 MIR2250 MIR2225
731-0717EA 4650 EUR
731-0717 731-0718 731-0716 MIRUMIR2225
Transfection reagents, transfection kit, TransIT®-mRNA
Transfektionsreagenzien
The TransIT®-mRNA Transfection Kit contains a high efficiency, low toxicity, large RNA transfection reagent for mammalian cells. Each kit is supplied with the TransIT-mRNA Transfection Reagent and the mRNA Boost Reagent.
  • Low cellular toxicity - maintain cell density and reduce experimental bias
  • High efficiency delivery - achieve RNA delivery in a large population of cells to ensure experimental success
  • Serum compatible - perform transfections in the presence of serum, which eliminates the need for a media change and maintains cellular health
  • Deliver various sizes of RNA - ideal for specialised applications, such as viral production and protein expression from mRNA