3-(Methoxycarbonyl)phenylboronsäurepinakolester
Lieferant: FLUOROCHEM
Synonyme:
3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid methyll ester, 2-(3-Methoxycarbonylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-(Methoxycarbonyl)phenylboronic acid Pinacol ester, Methyl 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate, 3-Methoxycarbonylphenylboronic acid pinacol ester
010812-5G
010812-1G
FLUO010812-5GEA
133
EUR
FLUO010812-5G
FLUO010812-1G
3-(Methoxycarbonyl)phenylboronsäurepinakolester
3-(Methoxycarbonyl)phenylboronsäurepinakolester
Formel:
C₁₄H₁₉BO₄ |
MDL:
MFCD03789258 CAS-Nummer: 480425-35-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...