3-(4-Fluorphenylcarbamoyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

BB-3088-1G BB-3088-5G
COBBBB-3088-1GEA 188 EUR
COBBBB-3088-1G COBBBB-3088-5G
3-(4-Fluorphenylcarbamoyl)phenylboronsäure
3-(4-Fluorphenylcarbamoyl)phenylboronsäure
Formel: C₁₃H₁₁BFNO₃
MDL: MFCD04115712
CAS-Nummer: 850567-35-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR