3-(4-Cyanphenylcarbamoyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (3-((4-Cyanophenyl)carbamoyl)phenyl)boronic acid

BB-3090-5G BB-3090-1G
COBBBB-3090-5GEA 451 EUR
COBBBB-3090-5G COBBBB-3090-1G
3-(4-Cyanphenylcarbamoyl)phenylboronsäure
3-(4-Cyanphenylcarbamoyl)phenylboronsäure
Formel: C₁₄H₁₁BN₂O₃
Molecular Weight: 266,0600 g/mol
MDL: MFCD06659886
CAS-Nummer: 850567-36-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR