3-(Cyclohexylcarbamoyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 3-(Cyclohexylaminocarbonyl)phenylboronic acid

BB-3065-1G BB-3065-5G BB-3065-25G
COBBBB-3065-1GEA 38.2 EUR
COBBBB-3065-1G COBBBB-3065-5G COBBBB-3065-25G
3-(Cyclohexylcarbamoyl)phenylboronsäure
3-(Cyclohexylcarbamoyl)phenylboronsäure
Formel: C₁₃H₁₈BNO₃
MDL: MFCD04115696
CAS-Nummer: 850567-25-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR