3-(Cyclopentylcarbamoyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (3-(Cyclopentylcarbamoyl)phenyl)boronic acid, 3-(Cyclopentylcarbamoyl)benzeneboronic acid

BB-3064-5G BB-3064-1G
COBBBB-3064-5GEA 0 EUR
COBBBB-3064-5G COBBBB-3064-1G
3-(Cyclopentylcarbamoyl)phenylboronsäure
3-(Cyclopentylcarbamoyl)phenylboronsäure
Formel: C₁₂H₁₆BNO₃
MDL: MFCD04115695
CAS-Nummer: 850567-24-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR