4-(Cyclopropylcarbamoyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 4-(Cyclopropylcarbamoyl)benzeneboronic acid, 4-(Cyclopropylaminocarbonyl)phenylboronic acid

BB-3013-5G BB-3013-1G
COBBBB-3013-5GEA 274 EUR
COBBBB-3013-5G COBBBB-3013-1G
4-(Cyclopropylcarbamoyl)phenylboronsäure
4-(Cyclopropylcarbamoyl)phenylboronsäure
Formel: C₁₀H₁₂BNO₃
Molecular Weight: 205,02 g/mol
MDL: MFCD03788419
CAS-Nummer: 515140-26-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR