3-Chlor-4-(cyclohexylcarbamoyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (3-Chloro-4-(cyclohexylcarbamoyl)phenyl)boronic acid

BB-2763-5G BB-2763-1G
COBBBB-2763-5GEA 451 EUR
COBBBB-2763-5G COBBBB-2763-1G
3-Chlor-4-(cyclohexylcarbamoyl)phenylboronsäure
3-Chlor-4-(cyclohexylcarbamoyl)phenylboronsäure
Formel: C₁₃H₁₇BClNO₃
Molecular Weight: 281.5400 g/mol
MDL: MFCD09475818
CAS-Nummer: 957034-65-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR