3-Chlor-4-(cyclopropylcarbamoyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (3-Chloro-4-(cyclopropylcarbamoyl)phenyl)boronic acid, 3-Chloro-4-(cyclopropylcarbamoyl)benzeneboronic acid

BB-2764-5G BB-2764-1G
COBBBB-2764-5GEA 1580 EUR
COBBBB-2764-5G COBBBB-2764-1G
3-Chlor-4-(cyclopropylcarbamoyl)phenylboronsäure
3-Chlor-4-(cyclopropylcarbamoyl)phenylboronsäure
Formel: C₁₀H₁₁BClNO₃
Molecular Weight: 239,46 g/mol
MDL: MFCD07363770
CAS-Nummer: 850589-44-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR