4-Chlor-3-(cyclopropylcarbamoyl)phenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
4-Chloro-3-(cyclopropylcarbamoyl)benzeneboronic acid, (4-Chloro-3-(cyclopropylcarbamoyl)phenyl)boronic acid
BB-2817-5G
BB-2817-1G
COBBBB-2817-5GEA
1580
EUR
COBBBB-2817-5G
COBBBB-2817-1G
4-Chlor-3-(cyclopropylcarbamoyl)phenylboronsäure
4-Chlor-3-(cyclopropylcarbamoyl)phenylboronsäure
Formel:
C₁₀H₁₁BClNO₃ Molecular Weight: 239.46 g/mol |
MDL:
MFCD07783865 CAS-Nummer: 871332-73-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...