3-(4-Fluorphenylcarbamoyl)phenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

PN-3088-5G PN-3088-1G
COBBPN-3088-5GEA 0 EUR
COBBPN-3088-5G COBBPN-3088-1G
3-(4-Fluorphenylcarbamoyl)phenylboronsäurepinakolester
3-(4-Fluorphenylcarbamoyl)phenylboronsäurepinakolester
Formel: C₁₉H₂₁BFNO₃
MDL: MFCD06659930
CAS-Nummer: 850567-58-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR