3-(Hydroxymethyl)benzonitril ≥97%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 3-Cyanobenzyl alcohol

H54212.03 H54212.06
H54212.03EA 37.2 EUR
H54212.03 H54212.06
3-(Hydroxymethyl)benzonitril ≥97%
3-(Hydroxymethyl)benzonitril

Purity: 97% CAS No.: 874-97-5 Molecular Formula: C8H7NO Formula Weight: 133.15 Flash Point: 109°(228°F)

Formel: C₈H₇NO
Molecular Weight: 133.15 g/mol
Flammpunkt: 109 °C (228 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD01321087
CAS-Nummer: 874-97-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR