3,5-Dimethyl-1H-pyrazol-4-boronsäurepinakolester, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
3,5-Dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole, 3,5-Dimethylpyrazole-4-boronic acid pinacol ester
M-Clarity™ Qualitätsstufe = MQ100
636010-1G
636010-5G
SIAL636010-1GEA
56.1
EUR
SIAL636010-1G
SIAL636010-5G
3,5-Dimethyl-1H-pyrazol-4-boronsäurepinakolester, Sigma-Aldrich®
3,5-Dimethyl-1H-pyrazol-4-boronsäurepinakolester
Formel:
C₁₁H₁₉BN₂O₂ Molecular Weight: 222,0918 g/mol |
MDL:
MFCD05663852 CAS-Nummer: 857530-80-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...