4-(N,N-Dimethylamino)phenylboronsäure 96%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 4-(Dimethylamino)benzeneboronic acid

359235000 359230025
ACRO359235000EA 44.4 EUR
ACRO359235000 ACRO359230025
4-(N,N-Dimethylamino)phenylboronsäure 96%
4-(N,N-Dimethylamino)phenylboronsäure
Formel: C₈H₁₂BNO₂
Molecular Weight: 165.00 g/mol
Schmelzpunkt: 227 °C
Lagertemperatur: Kühlschrank
MDL: MFCD01074642
CAS-Nummer: 28611-39-4

Bestellen

Spezifikation Testergebnisse

Appearance White to grey Powder and/or lumps
Infrared spectrum Conforms
Titration with NaOH ≥95 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR