4-(N,N-Dimethylamino)phenylboronsäure Hydrochlorid

Lieferant: COMBI-BLOCKS

Synonyme: 4-(N,N-Dimethylamino)Phenylboronic Acid Hydrochloride

BB-8464-5G BB-8464-1G
COBBBB-8464-5GEA 0 EUR
COBBBB-8464-5G COBBBB-8464-1G
4-(N,N-Dimethylamino)phenylboronsäure Hydrochlorid
4-(N,N-Dimethylamino)phenylboronsäure Hydrochlorid
Formel: C₈H₁₃BClNO₂
Molecular Weight: 201.4583 g/mol
MDL: MFCD04112547
CAS-Nummer: 1150114-73-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR