(1R,3S)-(+)-Camphersäure 99%

Lieferant: Thermo Scientific

Synonyme: D(+)-Camphoric acid, (1R,3S)-1,2,2-Trimethylcyclopentane-1,3-dicarboxylic acid, Levocarnitine Impurity B

108200050 108201000 108205000
ACRO108200050EA 25.7 EUR
ACRO108200050 ACRO108201000 ACRO108205000
(1R,3S)-(+)-Camphersäure 99%
(1R,3S)-(+)-Camphersäure
Formel: C₁₀H₁₆O₄
Molecular Weight: 200,23 g/mol
Schmelzpunkt: 183…187 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00001375
CAS-Nummer: 124-83-4
EINECS: 204-715-0

Bestellen

Spezifikation Testergebnisse

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 183 to 187 °C
Specific optical rotation +44.5° to +48.5° (20 °C, 589 nm) (c=10, C2H5OH)
Titration with NaOH ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR