(1R,3S)-(+)-Camphersäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: D(+)-Camphoric acid, (1R,3S)-1,2,2-Trimethylcyclopentane-1,3-dicarboxylic acid, Levocarnitine Impurity B

M-Clarity™ Qualitätsstufe = MQ200

C409-100G C409-5G
SIALC409-100GEA 95.3 EUR
SIALC409-100G SIALC409-5G
(1R,3S)-(+)-Camphersäure, Sigma-Aldrich®
(1R,3S)-(+)-Camphersäure
Formel: C₁₀H₁₆O₄
Molecular Weight: 200,23 g/mol
MDL: MFCD00001375
CAS-Nummer: 124-83-4
EINECS: 204-715-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR