4-(Cyclopropylcarbamoyl)phenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: N-Cyclopropyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide

PN-3013-5G PN-3013-1G
COBBPN-3013-5GEA 465 EUR
COBBPN-3013-5G COBBPN-3013-1G
4-(Cyclopropylcarbamoyl)phenylboronsäurepinakolester
4-(Cyclopropylcarbamoyl)phenylboronsäurepinakolester
Formel: C₁₆H₂₂BNO₃
Molecular Weight: 287,1618 g/mol
MDL: MFCD05663848
CAS-Nummer: 827614-68-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR