3-(Cyclohexylcarbamoyl)-4-fluorphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (3-(Cyclohexylcarbamoyl)-4-fluorophenyl)boronic acid

BB-2867-5G BB-2867-1G
COBBBB-2867-5GEA 911 EUR
COBBBB-2867-5G COBBBB-2867-1G
3-(Cyclohexylcarbamoyl)-4-fluorphenylboronsäure
3-(Cyclohexylcarbamoyl)-4-fluorphenylboronsäure
Formel: C₁₃H₁₇BFNO₃
Molecular Weight: 265,0900 g/mol
MDL: MFCD08235043
CAS-Nummer: 874219-24-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR