Methyl-8-brom-4-chlorchinolin-2-carboxylat

Lieferant: COMBI-BLOCKS

HC-2219-1G HC-2219-5G
COBBHC-2219-1GEA 98.3 EUR
COBBHC-2219-1G COBBHC-2219-5G
Methyl-8-brom-4-chlorchinolin-2-carboxylat
Methyl-8-brom-4-chlorchinolin-2-carboxylat
Formel: C₁₁H₇BrClNO₂
MDL: MFCD11504915
CAS-Nummer: 1072944-68-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR