Ethyl-4-chlorchinolin-3-carboxylat
Lieferant: BIONET RESEARCH
Synonyme:
4-Chloroquinoline-3-carboxylic acid ethyl ester, Ethyl 4-chloroquinoline-3-carboxylate
6J-051.500
6J-051.1
6J-051.5
6J-051.10
BIOT6J-051.500EA
0
EUR
BIOT6J-051.500
BIOT6J-051.1
BIOT6J-051.5
BIOT6J-051.10
Ethyl-4-chlorchinolin-3-carboxylat
Ethyl-4-chlorchinolin-3-carboxylat
Formel:
C₁₂H₁₀ClNO₂ Molecular Weight: 235.67 g/mol |
MDL:
MFCD00173407 CAS-Nummer: 13720-94-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...