Ethyl-8-chlor-4-hydroxychinolin-3-carboxylat

Lieferant: COMBI-BLOCKS

Synonyme: Ethyl 8-chloro-4-hydroxyquinoline-3-carboxylate, 8-Chloro-4-hydroxyquinoline-3-carboxylic acid ethyl ester

HC-2308-5G HC-2308-1G
COBBHC-2308-5GEA 362 EUR
COBBHC-2308-5G COBBHC-2308-1G
Ethyl-8-chlor-4-hydroxychinolin-3-carboxylat
Ethyl-8-chlor-4-hydroxychinolin-3-carboxylat
Formel: C₁₂H₁₀ClNO₃
Molecular Weight: 251,6657 g/mol
MDL: MFCD00024002
CAS-Nummer: 73987-37-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR