Ethyl-8-fluor-4-hydroxychinolin-3-carboxylat
Lieferant: COMBI-BLOCKS
Synonyme:
Ethyl 1,4-dihydro-8-fluor-4-oxoquinoline-3-carboxylat
PY-7379-1G
PY-7379-5G
COBBPY-7379-1GEA
25.9
EUR
COBBPY-7379-1G
COBBPY-7379-5G
Ethyl-8-fluor-4-hydroxychinolin-3-carboxylat
Ethyl-8-fluor-4-hydroxychinolin-3-carboxylat
Formel:
C₁₂H₁₀FNO₃ Molecular Weight: 235,211 g/mol |
MDL:
MFCD00052242 CAS-Nummer: 63010-69-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...