Ethyl-8-fluor-4-hydroxychinolin-3-carboxylat

Lieferant: Thermo Scientific

Synonyme: Ethyl 1,4-dihydro-8-fluor-4-oxoquinoline-3-carboxylat

BTB02003DA BTB02003EA BTB02003.50
MAYBBTB02003DAEA 0 EUR
MAYBBTB02003DA MAYBBTB02003EA MAYBBTB02003.50
Ethyl-8-fluor-4-hydroxychinolin-3-carboxylat
Ethyl-8-fluor-4-hydroxychinolin-3-carboxylat
Formel: C₁₂H₁₀FNO₃
Molecular Weight: 235,211 g/mol
MDL: MFCD00052242
CAS-Nummer: 63010-69-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR