Methyl-4-chlor-6-fluorchinolin-2-carboxylat

Lieferant: COMBI-BLOCKS

HC-2322-1G HC-2322-250M
COBBHC-2322-1GEA 595 EUR
COBBHC-2322-1G COBBHC-2322-250M
Methyl-4-chlor-6-fluorchinolin-2-carboxylat
Methyl-4-chlor-6-fluorchinolin-2-carboxylat
Formel: C₁₁H₇ClFNO₂
MDL: MFCD11855884
CAS-Nummer: 301823-61-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR