Ethyl-4-hydroxy-6-methoxychinolin-3-carboxylat
Lieferant: COMBI-BLOCKS
Synonyme:
4-Hydroxy-6-methoxyquinoline-3-carboxylic acid ethyl ester
HC-6349-1G
HC-6349-5G
COBBHC-6349-1GEA
98.7
EUR
COBBHC-6349-1G
COBBHC-6349-5G
Ethyl-4-hydroxy-6-methoxychinolin-3-carboxylat
Ethyl-4-hydroxy-6-methoxychinolin-3-carboxylat
Formel:
C₁₃H₁₃NO₄ Molecular Weight: 247.24 g/mol |
MDL:
MFCD00844092 CAS-Nummer: 77156-78-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...