Ethyl-4-hydroxy-7-methoxychinolin-3-carboxylat

Lieferant: COMBI-BLOCKS

Synonyme: Ethyl 4-hydroxy-7-methoxyquinoline-3-carboxylate

HC-6353-5G HC-6353-25G HC-6353-1G
COBBHC-6353-5GEA 451 EUR
COBBHC-6353-5G COBBHC-6353-25G COBBHC-6353-1G
Ethyl-4-hydroxy-7-methoxychinolin-3-carboxylat
Ethyl-4-hydroxy-7-methoxychinolin-3-carboxylat
Formel: C₁₃H₁₃NO₄
Molecular Weight: 247,2500 g/mol
MDL: MFCD00085986
CAS-Nummer: 63463-15-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR