Ethyl-4-hydroxy-8-methoxychinolin-3-carboxylat

Lieferant: COMBI-BLOCKS

Synonyme: Ethyl 4-hydroxy-8-methoxyquinoline-3-carboxylate, Ethyl 1,4-dihydro-8-methoxy-4-oxoquinoline-3-carboxylate

HC-6356-1G HC-6356-5G
COBBHC-6356-1GEA 102 EUR
COBBHC-6356-1G COBBHC-6356-5G
Ethyl-4-hydroxy-8-methoxychinolin-3-carboxylat
Ethyl-4-hydroxy-8-methoxychinolin-3-carboxylat
Formel: C₁₃H₁₃NO₄
Molecular Weight: 247,2466 g/mol
MDL: MFCD01829412
CAS-Nummer: 27568-04-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR