Ethyl-4-chlor-6-methylchinolin-3-carboxylat

Lieferant: COMBI-BLOCKS

HC-2758-5G HC-2758-1G
COBBHC-2758-5GEA 0 EUR
COBBHC-2758-5G COBBHC-2758-1G
Ethyl-4-chlor-6-methylchinolin-3-carboxylat
Ethyl-4-chlor-6-methylchinolin-3-carboxylat
Formel: C₁₃H₁₂ClNO₂
MDL: MFCD00173357
CAS-Nummer: 56824-87-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR