Methyl-4-chlor-8-methylchinolin-2-carboxylat

Lieferant: COMBI-BLOCKS

HC-2770-5G HC-2770-1G
COBBHC-2770-5GEA 303 EUR
COBBHC-2770-5G COBBHC-2770-1G
Methyl-4-chlor-8-methylchinolin-2-carboxylat
Methyl-4-chlor-8-methylchinolin-2-carboxylat
Formel: C₁₂H₁₀ClNO₂
MDL: MFCD12913953
CAS-Nummer: 1020101-33-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR