Methyl-7-chlor-4-hydroxy-8-methylchinolin-2-carboxylat
Lieferant: COMBI-BLOCKS
HC-6947-5G
HC-6947-1G
COBBHC-6947-5GEA
227
EUR
COBBHC-6947-5G
COBBHC-6947-1G
Methyl-7-chlor-4-hydroxy-8-methylchinolin-2-carboxylat
Methyl-7-chlor-4-hydroxy-8-methylchinolin-2-carboxylat
Formel:
C₁₂H₁₀ClNO₃ |
MDL:
MFCD11053779 CAS-Nummer: 1065074-49-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...