Tris(6,6,7,7,8,8,8-heptafluor-2,2-dimethyl-3,5-octandionato-O,O')europium(III) ≥99%, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Europium(III)-tris(1,1,1,2,2,3,3-heptafluoro-7,7-dimethyl-4,6-octanedionate), Eu(fod)3, Resolve-Al™ EuFOD, Europium-fod, Siever's reagent
160938-100MG
160938-1G
160938-5G
SIAL160938-100MGEA
23.2
EUR
SIAL160938-100MG
SIAL160938-1G
SIAL160938-5G
Tris(6,6,7,7,8,8,8-heptafluor-2,2-dimethyl-3,5-octandionato-O,O')europium(III) ≥99%, Sigma-Aldrich®
Tris(6,6,7,7,8,8,8-heptafluor-2,2-dimethyl-3,5-octandionato-O,O')europium(III)
Formel:
C₃₀H₃₀EuF₂₁O₆ Molecular Weight: 1037,49 g/mol |
MDL:
MFCD00064655 CAS-Nummer: 17631-68-4 EINECS: 241-616-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...