Tris(6,6,7,7,8,8,8-heptafluor-2,2-dimethyl-3,5-octandionato-O,O')ytterbium(III), Sigma-Aldrich®

Lieferant: Merck

Synonyme: Tris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionate)ytterbium(III), Yb(FOD)3, Tris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionato)ytterbium(III)

225835-1G 225835-5G
SIAL225835-1GEA 88 EUR
SIAL225835-1G SIAL225835-5G
Tris(6,6,7,7,8,8,8-heptafluor-2,2-dimethyl-3,5-octandionato-O,O')ytterbium(III), Sigma-Aldrich®
Tris(6,6,7,7,8,8,8-heptafluor-2,2-dimethyl-3,5-octandionato-O,O')ytterbium(III)
Formel: C₃₀H₃₀F₂₁O₆Yb
Molecular Weight: 1058.562 g/mol
MDL: MFCD00044289
CAS-Nummer: 18323-96-1
EINECS: 242-211-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR