(±)-Isoborneol 93% tech.

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: rel-(1R,2R,4R)-1,7,7-Trimethylbicyclo[2.2.1]heptan-2-ol

122490250 122495000
ACRO122490250EA 36.8 EUR
ACRO122490250 ACRO122495000
(±)-Isoborneol 93% tech.
(±)-Isoborneol
Formel: C₁₀H₁₈O
Molecular Weight: 154.2493 g/mol
Schmelzpunkt: 208…214 °C
Flammpunkt: 74 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00074821
CAS-Nummer: 124-76-5
EINECS: 204-712-4
UN: 1312
ADR: 4.1 III

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Powder or crystals and/or chunks
Infrared spectrum Conforms
GC ≥99.75 %
Density (20/4°C) 0.740 to 0.742
UV at 350 nm A: ≤0.01
Color scale ≤10 APHA
Residue after evaporation ≤0.0003 %
Peroxides ≤1 ppm (as H2O2)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR