(±)-Isoborneol ≥95%
Lieferant: Thermo Scientific
Synonyme:
rel-(1R,2R,4R)-1,7,7-Trimethylbicyclo[2.2.1]heptan-2-ol
B20632.14
B20632.22
B20632.36
B20632.14EA
21.3
EUR
B20632.14
B20632.22
B20632.36
(±)-Isoborneol ≥95%
(±)-Isoborneol
Purity: 95%
CAS No.: 124-76-5
Molecular Formula: C10H18O
Formula Weight: 154.25
Flash Point: 74°(165°F)
Odour: Strong
Beilstein Reference: 4126091
Merck Reference: 14,5128
UN No.: 1325
MDL No.: MFCD00074821
Formel:
C₁₀H₁₈O Molecular Weight: 154,2493 g/mol Schmelzpunkt: ∼210 °C (Subl.) Flammpunkt: 74 °C (165 °F) |
MDL:
MFCD00074821 CAS-Nummer: 124-76-5 EINECS: 204-712-4 ADR: , Merck Index: 14,5128 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...