(±)-Isoborneol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: rel-(1R,2R,4R)-1,7,7-Trimethylbicyclo[2.2.1]heptan-2-ol

M-Clarity™ Qualitätsstufe = MQ100

I13901-25G I13901-500G
SIALI13901-25GEA 47.9 EUR
SIALI13901-25G SIALI13901-500G
(±)-Isoborneol, Sigma-Aldrich®
(±)-Isoborneol
Formel: C₁₀H₁₈O
Molecular Weight: 154,2493 g/mol
MDL: MFCD00074821
CAS-Nummer: 124-76-5
EINECS: 204-712-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR