(±)-Isoborneol, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
rel-(1R,2R,4R)-1,7,7-Trimethylbicyclo[2.2.1]heptan-2-ol
M-Clarity™ Qualitätsstufe = MQ300
W215805-1KG-K
W215805-5KG-K
SIALW215805-1KG-KEA
154
EUR
SIALW215805-1KG-K
SIALW215805-5KG-K
(±)-Isoborneol, Sigma-Aldrich®
(±)-Isoborneol
Formel:
C₁₀H₁₈O Molecular Weight: 154,2493 g/mol |
MDL:
MFCD00074821 CAS-Nummer: 124-76-5 EINECS: 204-712-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...