2-(Tributylstannyl)oxazol 95%

Lieferant: Thermo Scientific
Gefahr

H55870.MD H55870.03 H55870.06
H55870.MDEA 71.6 EUR
H55870.MD H55870.03 H55870.06
2-(Tributylstannyl)oxazol 95%
2-(Tributylstannyl)oxazol

Purity: 97% CAS No.: 145214-05-7 Molecular Formula: C15H29NOSn Formula Weight: 358.11 Flash Point: 110°(230°F) Physical Form: Liquid Density: 1.71 Refractive Index: 1.4930 Appearance: Colourless Boiling Point: 108-110°/0.2mm UN No.: 2788 MDL No.: MFCD06798076

Formel: C₁₅H₂₉NOSn
Molecular Weight: 358,11 g/mol
Siedepunkt: 108…110 °C (0,2 mmHg)
Dichte: 1,71
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD06798076
CAS-Nummer: 145214-05-7
UN: 2788
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR