2-(Tributylstannyl)oxazol, Sigma-Aldrich®

Lieferant: Merck
Gefahr


M-Clarity™ Qualitätsstufe = MQ100

638617-1G 638617-5G
SIAL638617-1GEA 230 EUR
SIAL638617-1G SIAL638617-5G
2-(Tributylstannyl)oxazol, Sigma-Aldrich®
2-(Tributylstannyl)oxazol
Formel: C₁₅H₂₉NOSn
Molecular Weight: 358,11 g/mol
MDL: MFCD06798076
CAS-Nummer: 145214-05-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR