2-(Tributylstannyl)pyrazin

Lieferant: CHEMPUR
Gefahr

FL11062.POR 011062-1G 011062-5G
CHMPFL11062.POREA 0 EUR
CHMPFL11062.POR FLUO011062-1G FLUO011062-5G
2-(Tributylstannyl)pyrazin
2-(Tributylstannyl)pyrazin
Formel: C₁₆H₃₀N₂Sn
Molecular Weight: 369,12 g/mol
MDL: MFCD01319077
CAS-Nummer: 205371-27-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR