5-(Tributylstannyl)pyrimidin, Sigma-Aldrich®

Lieferant: Merck
Gefahr

719730-500MG 719730-1G 719730-5G
SIAL719730-500MGEA 69.4 EUR
SIAL719730-500MG SIAL719730-1G SIAL719730-5G
5-(Tributylstannyl)pyrimidin, Sigma-Aldrich®
5-(Tributylstannyl)pyrimidin
Formel: C₁₆H₃₀N₂Sn
Molecular Weight: 369,13 g/mol
MDL: MFCD01319056
CAS-Nummer: 144173-85-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR