1-Methyl-2-(tributylstannyl)pyrrol ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
H51438.03
H51438.06
H51438.03EA
96
EUR
H51438.03
H51438.06
1-Methyl-2-(tributylstannyl)pyrrol ≥98%
1-Methyl-2-(tributylstannyl)pyrrol
Purity: 98%
CAS No.: 118486-97-8
Molecular Formula: C17H33NSn
Formula Weight: 370.15
Flash Point: 98°(208°F)
Refractive Index: 1.511
UN No.: 2788
MDL No.: MFCD01319022
Formel:
C₁₇H₃₃NSn Molecular Weight: 370,15 g/mol Dichte: 1,122 Flammpunkt: 98 °C (208 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD01319022 CAS-Nummer: 118486-97-8 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...