2-(Tributylstannyl)thiazol, Sigma-Aldrich®

Lieferant: Merck
Gefahr


M-Clarity™ Qualitätsstufe = MQ100

642541-1G 642541-5G
SIAL642541-1GEA 145 EUR
SIAL642541-1G SIAL642541-5G
2-(Tributylstannyl)thiazol, Sigma-Aldrich®
2-(Tributylstannyl)thiazol
Formel: C₁₅H₂₉NSSn
Molecular Weight: 374,17 g/mol
MDL: MFCD01319057
CAS-Nummer: 121359-48-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR