1-(3-Chlor-5-(trifluormethyl)-2-pyridinyl)-1H-indol-3-carbaldehyd
Lieferant: BIONET RESEARCH
2H-989.500
2H-989.1
2H-989.5
2H-989.10
BIOT2H-989.500EA
0
EUR
BIOT2H-989.500
BIOT2H-989.1
BIOT2H-989.5
BIOT2H-989.10
1-(3-Chlor-5-(trifluormethyl)-2-pyridinyl)-1H-indol-3-carbaldehyd
1-(3-Chlor-5-(trifluormethyl)-2-pyridinyl)-1H-indol-3-carbaldehyd
Formel:
C₁₅H₈ClF₃N₂O |
MDL:
MFCD00664496 CAS-Nummer: 338770-32-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...