(S)-3,3'-Di(phenanthren-9-yl)-1,1'-binaphthyl-2,2'-diol

Lieferant: COMBI-BLOCKS

CS-9138-100M CS-9138-250M
COBBCS-9138-100MEA 0 EUR
COBBCS-9138-100M COBBCS-9138-250M
(S)-3,3'-Di(phenanthren-9-yl)-1,1'-binaphthyl-2,2'-diol
(S)-3,3'-Di(phenanthren-9-yl)-1,1'-binaphthyl-2,2'-diol
Formel: C₄₈H₃₀O₂
MDL: MFCD09027270
CAS-Nummer: 361342-50-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR