1-(3-Chlor-5-(trifluormethyl)-2-pyridinyl)tetrahydro-4(1H)-pyridinon

Lieferant: BIONET RESEARCH

Synonyme: 1-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]piperidin-4-one

8F-941.500 8F-941.1 8F-941.5 8F-941.10
BIOT8F-941.500EA 0 EUR
BIOT8F-941.500 BIOT8F-941.1 BIOT8F-941.5 BIOT8F-941.10
1-(3-Chlor-5-(trifluormethyl)-2-pyridinyl)tetrahydro-4(1H)-pyridinon
1-(3-Chlor-5-(trifluormethyl)-2-pyridinyl)tetrahydro-4(1H)-pyridinon
Formel: C₁₁H₁₀ClF₃N₂O
MDL: MFCD00231641
CAS-Nummer: 339029-35-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR