1-(3-Brom-5-methyl-2-pyridinyl)piperazin

Lieferant: COMBI-BLOCKS

PY-1115-5G PY-1115-1G
COBBPY-1115-5GEA 912 EUR
COBBPY-1115-5G COBBPY-1115-1G
1-(3-Brom-5-methyl-2-pyridinyl)piperazin
1-(3-Brom-5-methyl-2-pyridinyl)piperazin
Formel: C₁₀H₁₄BrN₃
MDL: MFCD12546612
CAS-Nummer: 1187386-35-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR