1-(3-Brom-5-methyl-2-pyridinyl)-4-propylpiperazin Hydrochlorid

Lieferant: COMBI-BLOCKS

PY-1172-5G PY-1172-1G PY-1172-250M
COBBPY-1172-5GEA 912 EUR
COBBPY-1172-5G COBBPY-1172-1G COBBPY-1172-250M
1-(3-Brom-5-methyl-2-pyridinyl)-4-propylpiperazin Hydrochlorid
1-(3-Brom-5-methyl-2-pyridinyl)-4-propylpiperazin Hydrochlorid
Formel: C₁₃H₂₁BrClN₃
MDL: MFCD12913992
CAS-Nummer: 1187386-03-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR