1-(3-Chlor-5-(trifluormethyl)-2-pyridinyl)-1,4-diazepan

Lieferant: FLUOROCHEM

Synonyme: 1-(3-Chloro-5-trifluoromethyl-2-pyridyl)homopiperazine

CFF206.0.5 CFF206.1 CFF206.2.5 017269-500MG 017269-1G 017269-2.5G
CHMPCFF206.0.5EA 348 EUR
CHMPCFF206.0.5 CHMPCFF206.1 CHMPCFF206.2.5 FLUO017269-500MG FLUO017269-1G FLUO017269-2.5G
1-(3-Chlor-5-(trifluormethyl)-2-pyridinyl)-1,4-diazepan
1-(3-Chlor-5-(trifluormethyl)-2-pyridinyl)-1,4-diazepan
Formel: C₁₁H₁₃ClF₃N₃
Molecular Weight: 279.69 g/mol
MDL: MFCD00238856
CAS-Nummer: 231953-40-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR